
5-Carboxytetramethylrhodamine (5-TAMRA) fluorescent probe
Catalog # AATB365
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:430.45 g/mol
ADR:5,II
:GHS07
CAS Number:91809-66-4
:MFCD1073638
Formula:C₂₅H₂₂N₂O₅
MDL Number:MFCD00269769