Fluo-8 AM
Supplier: AAT Bioquest
Jump to:
About this item
Calcium measurements are critical for numerous biological investigations.
- Can be loaded at rt
- Improved calcium response
- Significantly increased signal
- Suitable for all fluorescence platforms
Caution: For research use only.
- CAS Number:1345980-40-6
- Molecular Weight:1046.93 g/mol
- ADR:5,II
4 Options Available Below
Product Details & Documents
Calcium measurements are critical for numerous biological investigations.
- Can be loaded at rt
- Improved calcium response
- Significantly increased signal
- Suitable for all fluorescence platforms
Caution: For research use only.
- CAS Number:1345980-40-6
- Molecular Weight:1046.93 g/mol
- ADR:5,II



