Order Entry
United Kingdom
ContactUsLinkComponent
Resorufin benzyl ether ≥90%
Irritant
Resorufin benzyl ether ≥90%
Catalog # AATB15020
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Resorufin benzyl ether ≥90%
Catalog # AATB15020
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
ADR:5,II
 :303.31 g/mol
 :GHS07
CAS Number:87687-02-3
Formula:C₁₉H₁₃NO₃
MDL Number:MFCD00171613