Order Entry
United Kingdom
ContactUsLinkComponent
Disinfectant, Anioxyde 1000 LD
Disinfectant, Anioxyde 1000 LD
  DMDA2824299EV
Disinfectant, Anioxyde 1000 LD
  DMDA2824299EV
  • Capacity:
    5 L
  • Description:
    Anioxyde 1000 LD with integrated activator
  • Environmentally Preferable:
  • Packed:
    Canister
  • Pk:
    4
High-level disinfectant for thermosensitive instruments and endoscopy equipment.
  • Shelf life before use: 24 months
  • High level disinfectant from PHERA® system, active against spores of bacteria
  • Extemporaneous production of peracetic acid
  • Alternative to glutaraldehyde
  • Aldehyde free: no fixation of proteins
  • Possible control of bath efficacy with test-strips
  • Total effectiveness in 5 minutes
 : Use biocides safely. Always read the label and product information before use.
For professional use only.