


1H,1H,2H,2H-Heptadecafluorodecyl acrylate, colourless, liquid
EHERCA15986595
:
- Pk:100 mg
- Seq:2440
- Accurate mass:518.0175
- Storage temperature:4 °C
- Shipping temperature:20 °C
- Odour:Odourless
:
:518.17 g/mol
:90 °C
:132 °C
:3082
:9,III
:GHS07
:GHS08
:GHS09
:27905-45-9
:C₁₃H₇F₁₇O₂
:MFCD00042306