Claritas PPT® Multi-element Standards Solution 1 for ICP and ICP-MS Instrumentation
Catalog # COLE04568-80
Supplier: Cole-Parmer
CAS Number:
Specifications
- Concentration:10 µg/ml
- Pk:125 mL
- Description:Claritas PPT® Grade ICP-MS multi-element solution 1, 10 µg/ml in 5% HNO₃
- Environmentally Preferable:
- UN No.:3264
- Shelf life:12 Months
- Physical form:Liquid
Specifications
About this item
Calibrate ICP-MS instrumentation prior to analysis.
- Dilute to the concentration appropriate for the instrument
- Multi-element internal standards in 5% HNO₃ matrix
Multi element standards for Cerium, Dysprosium, Erbium, Europium,Gadolinium, Holmium, LaLutetium, Neodymium, Praseodymium, Samarium, Scandium, Terbium, Thulium, Ytterbium, Yttrium.
Certifications: Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance.