Recombinant Mouse CD40LG (from Baculovirus), His-tag
Catalog # ABNOP9075.5UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:5 µG
- Protein/peptide type:Recombinant
- Source:Baculovirus
- Species:Mouse
- Environmentally Preferable:
- Tag sequence:His-tag
- Gene ID:P27548
- Protein/peptide name:CD40LG
- Sequence:112a.a.-260a.a.