Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Mouse RETN (from E. coli), Flag-tag
Recombinant Mouse RETN (from E. coli), Flag-tag
ABNOP9161.25UG
:
Abnova
:
Recombinant Mouse RETN (from E. coli), Flag-tag
ABNOP9161.25UG
:
Abnova
:
Pk:
25 µG
Protein/peptide type:
Recombinant
Source:
E. coli
Species:
Mouse
Environmentally Preferable:
Tag sequence:
Flag-tag
Protein/peptide name:
RETN