Order Entry
United Kingdom
ContactUsLinkComponent
Recombinant Mouse IL15 (from E. coli), His-tag
Recombinant Mouse IL15 (from E. coli), His-tag
  ABNOP8348.10UG
 :  Abnova
 :  
Recombinant Mouse IL15 (from E. coli), His-tag
  ABNOP8348.10UG
 :  Abnova
 :  

 

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Environmentally Preferable:
  • Tag sequence:
    His-tag
  • Gene ID:
    P48346
  • Protein/peptide name:
    IL15
  • Sequence:
    49a.a.-162a.a.