Order Entry
United Kingdom
ContactUsLinkComponent
 
Recombinant Human FGF23 (from E. coli), His-tag
  ABNOP8638.10UG
 :  Abnova
 :  
Recombinant Human FGF23 (from E. coli), His-tag
  ABNOP8638.10UG
 :  Abnova
 :  

 

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Environmentally Preferable:
  • Tag sequence:
    His-tag
  • Protein/peptide name:
    FGF23