Order Entry
United Kingdom
ContactUsLinkComponent
Recombinant Mouse IL2 (from E. coli)
Recombinant Mouse IL2 (from E. coli)
Catalog # ABNOP8244.20UG
Supplier:  Abnova
CAS Number:  
Recombinant Mouse IL2 (from E. coli)
Catalog # ABNOP8244.20UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Environmentally Preferable:
  • Gene ID:
    P04351
  • Protein/peptide name:
    IL2
  • Sequence:
    22a.a.-169a.a.

Specifications

About this item