Recombinant Mouse GDA (from E. coli), His-tag
Catalog # ABNOP8027.20UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:20 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Tag sequence:His-tag
- Gene ID:Q9R111
- Protein/peptide name:GDA
- Sequence:1a.a.-454a.a.