Recombinant GLDA (from E. coli), His-tag
Catalog # ABNOP7936.100UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:100 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:
- Environmentally Preferable:
- Tag sequence:His-tag
- Gene ID:P0A9S5
- Protein/peptide name:GLDA
- Sequence:1a.a.-367a.a.