Order Entry
United Kingdom
ContactUsLinkComponent
Anti-H3-4 Rabbit Recombinant Antibody [clone: HISH3S28-D6] (APC)
Anti-H3-4 Rabbit Recombinant Antibody [clone: HISH3S28-D6] (APC)
Catalog # ABNORAB02904.100
Supplier:  Abnova
Anti-H3-4 Rabbit Recombinant Antibody [clone: HISH3S28-D6] (APC)
Catalog # ABNORAB02904.100
Supplier:  Abnova

Specifications

  • Antigen symbol:
    H3-4
  • Clonality:
    Recombinant
  • Clone:
    HISH3S28-D6
  • Conjugation:
    APC (Allophycocyanin)
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human H3-4.
  • Pk:
    100 AU

Specifications

About this item