Anti-DISC1 Rabbit Recombinant Antibody [clone: R03-7F5]
Catalog # ABNORAB02322.100UL
Supplier: Abnova
Specifications
- Antigen symbol:DISC1
- Clonality:Recombinant
- Clone:R03-7F5
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Immunogen:Recombinant rabbit monoclonal antibody raised against human DISC1.
- Pk:100 µl