Anti-DDX5 Rabbit Recombinant Antibody [clone: R04-2K2]
Catalog # ABNORAB02326.100UL
Supplier: Abnova
Specifications
- Antigen symbol:DDX5
- Clonality:Recombinant
- Clone:R04-2K2
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Immunogen:Recombinant rabbit monoclonal antibody raised against human DDX5.
- Pk:100 µl