Anti-TNFRSF10D Rabbit Recombinant Antibody [clone: R01-1F0]
Catalog # ABNORAB02329.100UL
Supplier: Abnova
Specifications
- Antigen symbol:TNFRSF10D
- Clonality:Recombinant
- Clone:R01-1F0
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Immunogen:Recombinant rabbit monoclonal antibody raised against human TNFRSF10D.
- Pk:100 µl