Anti-BCMA (Met1-Ala54) HEK293 cells Recombinant Antibody [clone: DM3]
ABNORAB01763.10UL
: Abnova
- Antigen symbol:BCMA (Met1-Ala54)
- Clonality:Recombinant
- Clone:DM3
- Host:HEK293
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Immunogen:Recombinant rabbit monoclonal antibody raised against human BCMA (Met1-Ala54) produced by using human HEK293 cells
- Pk:10 µl