£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United Kingdom
ContactUsLinkComponent
Antibodies
Anti-UBB Rabbit Recombinant Antibody [clone: R01-7F1]
Antigen symbol:
UBB
Clonality:
Recombinant
Clone:
R01-7F1
Host:
Rabbit
Isotype:
Reactivity:
Human
Environmentally Preferable:
Immunogen:
Recombinant rabbit monoclonal antibody raised against human UBB.
Pk:
100 µl