Anti-CDK1 Rabbit Recombinant Antibody [clone: R05-6I8]
Catalog # ABNORAB01516.100UL
Supplier: Abnova
Specifications
- Antigen symbol:CDK1
- Clonality:Recombinant
- Clone:R05-6I8
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Immunogen:Recombinant rabbit monoclonal antibody raised against synthetic peptide of human CDK1.
- Pk:100 µl