Anti-SEMA4D Rabbit Recombinant Antibody [clone: R05-2E3]
Catalog # ABNORAB01216.100UL
Supplier: Abnova
Specifications
- Antigen symbol:SEMA4D
- Clonality:Recombinant
- Clone:R05-2E3
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Immunogen:Recombinant rabbit monoclonal antibody raised against human SEMA4D.
- Pk:100 µl