Anti-CREB1 Rabbit Recombinant Antibody [clone: R07-3C4]
Catalog # ABNORAB01231.100UL
Supplier: Abnova
Specifications
- Antigen symbol:CREB1
- Clonality:Recombinant
- Clone:R07-3C4
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Immunogen:Recombinant rabbit monoclonal antibody raised against human CREB1.
- Pk:100 µl