Order Entry
United Kingdom
ContactUsLinkComponent
Anti-VIM Rabbit Recombinant Antibody [clone: R02-8C4]
Anti-VIM Rabbit Recombinant Antibody [clone: R02-8C4]
Catalog # ABNORAB01197.100UL
Supplier:  Abnova
Anti-VIM Rabbit Recombinant Antibody [clone: R02-8C4]
Catalog # ABNORAB01197.100UL
Supplier:  Abnova

Specifications

  • Antigen symbol:
    VIM
  • Clonality:
    Recombinant
  • Clone:
    R02-8C4
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Immunogen:
    Recombinant rabbit monoclonal antibody raised against human VIM.
  • Pk:
    100 µl

Specifications

About this item