Condenser, C-ABBE 90
NIKOMBL71105
: NIKON
- Description:Condernser, C-ABBE 90
- Environmentally Preferable:
- Pk:1
C-AB Abbe condenser for biological microscopies.
- Ext.dimension: 1,90 mm
- Magnification: 4 to 100X
- Weight: 90 g
C-AB Abbe condenser for biological microscopies.