Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Eclipse Ci-L plus Upright Microscope
Eclipse Ci-L plus Upright Microscope
Upright microscope, Ci-L Plus, Binocular
Catalog # NIKOMBA95010
Supplier:   NIKON
Eclipse Ci-L plus Upright Microscope
Catalog # NIKOMBA95010
Supplier:  NIKON
Eclipse Ci-L plus Upright Microscope
Upright microscope, Ci-L Plus, Binocular
Price
£1,900.00
Log in for availability
1 * 1 items
Product Details & Documents
The Nikon ECLIPSE Ci-L plus is a biological microscope designed with a focus on ease-of-use as well as the health and working styles of people involved in research.
  • Ergonomic designed with attention toward eliminating neck, shoulder and back strain
  • Maintains optimal brightness with the light intensity management (LIM)
  • Keeps the same observation posture with automatic brightness adjustment
  • Status display at base of the microscope for easy confirmation at a glance without changing your observation posture
  • Turning the nosepiece automatically adjusts the scale bar
The eyepiece tube can be inclined and extended, and the angle of the elbow can be adjusted by changing the height of the handle, so that you can always adopt a posture that suits you, thereby reducing physical strain.
The light intensity management (LIM) function developed by Nikon automatically stores any changes to the brightness settings. This helps avoid drastic changes in brightness when switching between different magnification during observations, thereby helping mitigate eye strain.
Specifications
  • Model:
    Ci-L Plus
  • Description:
    Upright microscope
  • Type:
    Binocular
  • Focusing system:
    Coaxial coarse and fine focusing system with tension adjustment
  • Cat. No.:
    NIKOMBA95010
  • Observation method:
    Brightfield, darkfield, phase contrast and polarisation
  • Condenser:
    C-AB Abbe condenser
  • Nosepiece:
    Sextuple
  • Illumination:
    High luminescent white LED
  • Stage:
    Cross travel 78×54 mm, with vernier calibrations, stage handle height and torque adjustable for all stages
  • Electrical:
    5 W
  • Weight:
    13,3 kg