Digital Microscope, DVM6 M
Product Details & Documents
The DVM6 digital microscope solution is fast, reliable, and easy to use - no matter if you work in quality assurance, failure analysis, research and development, or forensics.
- The DVM6 works straightforward and saves you time inspecting various samples
- This solution enables users to start working with minimal training requirements
If the sample is too large or bulky for the standard DVM6 configurations, the modular DVM6 M offers you more flexibility for a wide range of applications. With a special interface adapter, the DVM6 M zoom module can be used with selected stands from the Leica M series stereo microscopes for large (max. stage travel range: 150×100 mm) and tall (up to 340 mm) samples.
The DVM6 digital microscope is a versatile solution that helps you gain insights during your sample investigations, stay focused on your work thanks to a convenient operation, and improve reliability of results with reproducible imaging conditions.
Specifications
- Model:DVM6 M
- Description:Digital microscope zoom module package
- Additional features:Zoom module with 16:1 zoom and integrated camera, data connection via USB3, interface module required for mounting on focusing column
Accessories
Product Information
- Catalog #AvailabilityPrice
- Manual IsoPro XY stage, 6×4"
Product Information
- Catalog #AvailabilityPrice
- Scanning stage adapter