- Description:Safety storage cabinet ION-CORE-90 STORE model IO90.195.120.CS.WDC without interior equipment body colour anthracite grey (RAL 7016) and wing doors in gentian blue (RAL 5010)
- W×D×H:1193×615×1953 mm
- Pk:1
- Weight:424 kg
- W×D×H int.:1050×522×1647 mm
Safe passive storage of lithium-ion batteries.
- Robust construction and longevity
- Convenient cabinet entry
- Integrated potential-free alarm contact
- No unauthorised use
- Integrated transport base
- Easy alignment
- Smoke detector
- Pressure relief
- All-around protection
Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated textured surface. Convenient cabinet entry: open both doors at the same time with just one hand. Integrated potential-free alarm contact for strongly recommended alarm forwarding. Optionally available with module for remote signalling.
No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green). Permanent locking of the doors by automatic 2-point locking. Integrated transport base: In the event of an emergency, immediate evacuation and transport of the cabinet to a safe place outdoors.
Easy alignment: adjusting aids to compensate for uneven floor. Smoke detector for fire detection and forwarding the alarm to the building management system. Pressure relief in the head section.
All-around protection: 90 minutes fire protection from outside to inside (type 90 / type tested in accordance with EN 14470-1) and for more than 90 minutes fire resistance for fires from inside to outside.