Kinesis Non Sterile Syringe Filters, Nylon
Hydrophilic nylon membranes exhibit strong mechanical stability and medium protein binding.
- Use with aqueous and alcoholic solutions and most HPLC solvents
- Solvent-resistant filter housing with nominal extractable
- Leak-free Luer-Lok® and Luer-slip connections
- Color-coded filters for easy organization
Prepare reliable aqueous, organic, and biomolecular samples with membrane filtration inside a polypropylene (PP) housing. Each housing comes with a color-coded retainer ring seal to prevent leaks and sample loss, and the color helps to visually show you the membrane type inside the housing.
Syringe filters have Luer-Lok and Luer-slip connections for easy, quick and tight connections to tubing, syringes, or needles. Filter packs are available in a resealable storage container to enhance cleanliness during multiple opening and closings.
- Catalog No:
- COLEESF-NY-30-045D
- COLEESF-NY-30-022D
- Max. operating pressure:
- 100 psi
- 100 psi
Frequently Bought Together