Sodium hydrogen carbonate 99.0-100.5% (by acidimetry) USP, BP, Ph. Eur., pure, Pharma Grade
Supplier: PanReac AppliChem
Certificates
About this item
CAS Number:144-55-8
:
Formula:NaHCO₃
MDL Number:MFCD00003528
:84.01 g/mol
Storage Temperature:Ambient
EINECS:205-633-8
Specification Test Results
- Assay (Acidim.):99.0-100.5%
- Identity according to Pharmacopoeias::passes test
- Appearance of solution:passes test
- Loss on drying:0.25%
- Chloride (Cl):0.005%
- Sulfur compounds (as SO4):0.015 %
- Ammonium (NH4):0.002%
- Phosphate (PO4):0.002 %
- Sulfate (SO4):0.01%
- Carbonate:passes test
- Residual solvents (Ph.Eur/USP):passes test
- As:0.00015 %
- Ca:0.01 %
- Fe:0.0005 %
Specification Test Results
Frequently Bought Together