Mineral oil light FCC, low viscosity
:8042-47-5
:
:MFCD00131611
:Ambient
- Carbon Number at 5% Distillation Point:17 - 25
- Specific Gravity @ 25°C:0.829 - 0.859
- Ultraviolet Absorbance (Polynuclear Hydrocabons):To Pass Test
- Viscosity @ 100°C:3 - 11 cSt
- Lead (Pb):max. 1 mg/kg
Frequently Bought Together