Calcium phosphate tribasic, powder NF
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:12167-74-7
Formula:Ca₅HO₁₃P₃
MDL Number:MFCD00010904
:502.306 g/mol
Storage Temperature:Ambient
Specification Test Results
- Assay (Ca):34.0 - 40.0%
- Loss On Ignition:max. 8.0 %
- Water-Soluble Substances:max. 0.5 %
- Acid-Insoluble Substances:max. 0.2 %
- Carbonate (CO₃):To Pass Test
Specification Test Results
Frequently Bought Together