Specifications
- Antibody type:Primary
- Antigen name:ASB17
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:81-180/295
- Form:Liquid
- Gene ID:127247
- Antigen synonyms:ASB-17|Ankyrin repeat and SOCS box containing 17|ASB17_HUMAN|Ankyrin repeat and SOCS box protein 17|ASB17
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human ASB17
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Members of the suppressor of cytokine signaling (SOCS) family of proteins contain C-terminal regions of homology called the SOCS box, which serves to couple SOCS proteins and their binding partners with the Elongin B and C complex. Several other families of proteins also contain SOCS boxes, but differ from the SOCS proteins in the type of domain they contain upstream of the SOCS box. The largest family of SOCS box-containing proteins is the ankyrin repeat and SOCS box-containing (ASB) protein family. Members of the ASB family include ASB-1 through ASB-18 and are involved in a variety of biological processes. ASB-17 is a 295 amino acid member of this family. ASB-17 contains one ankyrin repeat and one SOCS box domain. ASB-17 is thought to be a substrate-recognition subunit of a SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin protein ligase complex. This complex mediates the ubiquitination of target proteins and their subsequent proteasomal degradation.
Type: Primary
Antigen: ASB17
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 81-180/295
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat