Anti-BRSK1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-7905R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:BRSK1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Epitope:101-200/778
- Form:Liquid
- Gene ID:84446
- Antigen synonyms:Protein kinase SAD1A|SAD1|BRSK1_HUMAN|FLJ43009|Brsk1|SAD1A|BR serine/threonine protein kinase 1|KIAA1811|Serine/threonine-protein kinase SAD-B|BR serine/threonine-protein kinase 1|Synapses of Amphids Defective homolog|SadB kinase short isoform|Brain-specic serine/threonine-protein kinase 1|Serine/threonine kinase SAD B|SAD1 kinase|SADB|BR serine/threonine kinase 1|SAD 1|BRSK 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:86 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human BRSK1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Required for the polarisation of forebrain neurons which endows axons and dendrites with distinct properties, possibly by locally regulating phosphorylation of microtubule-associated proteins (By similarity). May be involved in the regulation of G2/M arrest in response to UV- or methyl methane sulfonate (MMS)-induced, but not IR-induced, DNA damage. Phosphorylates WEE1 and CDC25B in vitro and CDC25C in vitro and in vivo.Tissue specificity: Widely expressed, with highest levels in brain and testis. Protein levels remain constant throughout the cell cycle.
Type: Primary
Antigen: BRSK1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/778
Host: Rabbit
Isotype: IgG
Reactivity: Rat