Anti-CMG1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-7050R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:CMG1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Epitope:51-150/600
- Form:Liquid
- Gene ID:80173
- Antigen synonyms:intraflagellar transport 74 homolog|Intraflagellar transport protein 74 homolog|CMG-1|IFT74_HUMAN|Ccdc2|Coiled-coil domain containing 2|Coiled-coil domain-containing protein 2|CMG 1|Capillary morphogenesis gene 1 protein|IFT74|Capillary morphogenesis protein 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:69 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human CMG1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
No data available.
Type: Primary
Antigen: CMG1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/600
Host: Rabbit
Isotype: IgG
Reactivity: Human