Order Entry
United Kingdom
ContactUsLinkComponent
Estrogen (Human) ELISA Kit
Estrogen (Human) ELISA Kit
Catalog # ABCAAB285239-96
Supplier:  Abcam
CAS Number:  
undefined
Estrogen (Human) ELISA Kit
Catalog # ABCAAB285239-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay Type:
    Competitive
  • Host:
  • Primary antibody reactivity:
    Human
  • Description:
    Estrogen (human) ELISA kit
  • Environmentally Preferable:
  • Tests per kit:
    100 assays

Specifications

About this item

An competitive ELISA kit to quantify Estrogen in human serum, plasma and other biological fluids.

  • Easy, convenient and time-saving method to measure the level of Estrogen in human serum, plasma, tissue homogenates, culture supernatants and other biological fluids
  • Detection Range: 15,6 - 1000 pg/ml
  • Sensitivity: <9,375 pg/ml

Estrogen is the primary female sex hormone. It is responsible for the development and regulation of the female reproductive system and secondary sex characteristics. Estrogen may also refer to any substance, natural or synthetic, that mimics the effects of the natural hormone. The steroid 17β-estradiol is the most potent and prevalent endogenous estrogen, although several metabolites of estradiol also have estrogenic hormonal activity. Synthetic estrogens are used as part of some oral contraceptives, in estrogen replacement therapy for postmenopausal women, and in hormone replacement therapy for trans women. BioVision’s Estrogen ELISA kit is a Competitive ELISA assay for the quantitative measurement of human Estrogen in serum, plasma and cell culture supernatants. The density of color is proportional to the amount of human Estrogen captured from the samples.