Order Entry
United Kingdom
ContactUsLinkComponent
PLAC1 Overexpression Lysate (Adult Normal)
PLAC1 Overexpression Lysate (Adult Normal)
Catalog # NOVUNBL1-14479
Supplier:  Novus Biologicals
CAS Number:  
undefined
PLAC1 Overexpression Lysate (Adult Normal)
Catalog # NOVUNBL1-14479
Supplier:  Novus Biologicals
CAS Number:  

Specifications

  • Cell Format:
    Cryopreserved
  • Pk:
    100 µG
  • Environmentally Preferable:
  • Tested applications:
    WB

Specifications

About this item

PLAC1 Overexpression Lysate (Adult Normal)

Tested Applications: Western Blot.