Anti-ALS2CR1/N3L1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-11649R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:ALS2CR1/N3L1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:201-300/377
- Form:Liquid
- Antigen synonyms:N3 NGG1 interacting factor 3-like 1|N3-like protein 1|N3 NGG1 interacting factor 3-like 1 S. pombe|N3L1|Ngg1 interacting factor 3-like 1|ALS2CR1|Amyotrophic lateral sclerosis 2 juvenile chromosome region, candidate 1|N3 Ngg1 interacting factor 3, S.pombe homolog-like 1|Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 1 protein|MDS015|N3L_HUMAN|CALS-7
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:42 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human ALS2CR1/NIF3L1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
The NGG1 interacting factor 3-like 1 (NIF3L1) is a 377 amino acid protein expressed mainly in the cytoplasm of cells in several different tissues. It has been highly conserved throughout evolution, from bacteria to mammals. NIF3L participates in retinoic acid-primed neural differentiation of P19 embryonic carcinoma cells by cooperating with Trip15/CSN2, a transcriptional corepressor/component of COP9 signalosome. NIF3L1 interacts with itself and with the NIF3L1 binding protein 1 (NIF3L1 BP1), which is a novel protein presumed to contain a leucine zipper domain.
Type: Primary
Antigen: ALS2CR1/N3L1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/377
Host: Rabbit
Isotype: IgG
Reactivity: