Specifications
- Antibody type:Primary
- Antigen name:GIOT1/ZNF461
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:191-240/563
- Form:Liquid
- Gene ID:92283
- Antigen synonyms:ZNF461|Zinc finger protein 461|Gonadotropin inducible transcription repressor 1|Gonadotropin-inducible ovary transcription repressor 1|ZN461_HUMAN|GIOT-1|Gonadotropin inducible ovary transcription repressor 1|GIOT 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:66 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GIOT1/ZNF461 (191-240aa)
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
GIOT-1 is a 563 amino acid protein belonging to the Kr_ppel C2H2-type zinc-finger protein family. localised to the nucleus, GIOT-1 is widely expressed in tissues, with highest levels in liver, kidney, small intestine, pancreas and thymus. GIOT-1 contains 12 C2H2-type zinc fincers and one KRAB domain. Because the KRAB domain functions as a transcriptional repressor when attached to the template DNA, GIOT-1 is thought to be involved in transcriptional regulation. The gene encoding GIOT-1 is localised to chromosome 19q13.12 and two isoforms of GIOT-1 exist as a result of alternative splicing events.
Type: Primary
Antigen: GIOT1/ZNF461
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 191-240/563
Host: Rabbit
Isotype: IgG
Reactivity: Mouse