Anti-RFC2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-11305R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:RFC2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:281-354/354
- Form:Liquid
- Gene ID:5982
- Antigen synonyms:A1 40 kDa subunit|Replication factor C subunit 2|Replication factor C 40 kDa subunit|Activator 1 subunit 2|RF-C 40 kDa subunit|RFC40|RFC2|Activator 1 40 kDa subunit
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:39 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human RFC2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
The elongation of primed DNA templates by DNA polymerase delta and epsilon requires the action of the accessory proteins proliferating cell nuclear antigen (PCNA) and activator 1. This subunit binds ATP (By similarity).
Type: Primary
Antigen: RFC2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 281-354/354
Host: Rabbit
Isotype: IgG
Reactivity: Mouse