- Antibody type:Primary
- Antigen name:Microcephalin 1/BRIT1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:11-110/835
- Form:Liquid
- Antigen synonyms:BRCT repeat inhibitor of TERT expression 1|Microcephaly primary autosomal recessive 1|MCPH 1|BRIT 1|Microcephalin-1|MCT antibody Microcephalin 1|MCPH1_HUMAN|FLJ12847|MCPH1|Hypothetical protein FLJ12847
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:93 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Microcephalin 1/BRIT1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Microcephalin modulates brain size and has been proliferating under strong positive selection for several thousand years, although the nature of the positive selection is poorly understood. Human Microcephalin contains three BRCA1 C-terminal (BRCT) domains and shares 57% identity with its mouse ortholog, the most conserved regions being BRCT domains where there is 80% identity. Predominant expression of human Microcephalin is observed in fetal brain, liver and kidney tissues and is expressed during neurogenesis in mice. Microcephalin displays significantly higher rates of protein evolution in primates than in rodents; this trend is most noticeable for the subset of genes associated with nervous system development. Microcephalin has a very young, single nucleotide, polymorphism haplotype associated with modern humans; this gene is presumably still evolving in Homo sapiens. It functions in DNA damage response and regulation of cell cycle checkpoints.
Type: Primary
Antigen: Microcephalin 1/BRIT1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 11-110/835
Host: Rabbit
Isotype: IgG
Reactivity: Mouse