Order Entry
United Kingdom
ContactUsLinkComponent
Anti-GLUR3/GRIA 3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-GLUR3/GRIA 3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-1799R-A680
Supplier:  Bioss
Anti-GLUR3/GRIA 3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-1799R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    GLUR3/GRIA 3
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Epitope:
    451-550/894
  • Form:
    Liquid
  • Gene ID:
    2892
  • Antigen synonyms:
    GluR-3|GluA3|GLUR3|AMPA-selective glutamate receptor 3|GLURC|Glutamate receptor ionotropic, AMPA 3|GLUR-K3|GRIA3|GLUR-C|MRX94|Glutamate receptor 3
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    98 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human GLUR3
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Receptor for glutamate that functions as ligand-gated ion channel in the central nervous system and plays an important role in excitatory synaptic transmission. L-glutamate acts as an excitatory neurotransmitter at many synapses in the central nervous system. Binding of the excitatory neurotransmitter L-glutamate induces a conformation change, leading to the opening of the cation channel, and thereby converts the chemical signal to an electrical impulse. The receptor then desensitizes rapidly and enters a transient inactive state, characterised by the presence of bound agonist. In the presence of CACNG4 or CACNG7 or CACNG8, shows resensitization which is characterised by a delayed accumulation of current flux upon continued application of glutamate.

Type: Primary
Antigen: GLUR3/GRIA 3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 451-550/894
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat