Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-ATF4/CREB-2 Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-1531R-CY7
 :  Bioss
Anti-ATF4/CREB-2 Rabbit Polyclonal Antibody (Cy7®)
  BOSSBS-1531R-CY7
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    ATF4/CREB-2
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rabbit,
    Human,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    251-351/351
  • Form:
    Liquid
  • Gene ID:
    468
  • Antigen synonyms:
    DNA-binding protein TAXREB67|TAXREB67|cAMP-dependent transcription factor ATF-4|Cyclic AMP-dependent transcription factor ATF-4|TXREB|Tax-responsive enhancer element-binding protein 67|CREB2|Activating transcription factor 4|cAMP-responsive element-binding protein 2|CREB-2|Cyclic AMP-responsive element-binding protein 2|ATF4
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    38 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human ATF4
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Transcriptional activator. Binds the cAMP response element (CRE) (consensus: 5'-GTGACGT[AC][AG]-3'), a sequence present in many viral and cellular promoters. Cooperates with FOXO1 in osteoblasts to regulate glucose homeostasis through suppression of beta-cell production and decrease in insulin production (By similarity). It binds to a Tax-responsive enhancer element in the long terminal repeat of HTLV-I. Regulates the induction of DDIT3/CHOP and asparagine synthetase (ASNS) in response to ER stress. In concert with DDIT3/CHOP, activates the transcription of TRIB3 and promotes ER stress-induced neuronal apoptosis by regulating the transcriptional induction of BBC3/PUMA. Activates transcription of SIRT4. Regulates the circadian expression of the core clock component PER2 and the serotonin transporter SLC6A4. Binds in a circadian time-dependent manner to the cAMP response elements (CRE) in the SLC6A4 and PER2 promoters and periodically activates the transcription of these genes.

Type: Primary
Antigen: ATF4/CREB-2
Clonality: Polyclonal
Clone:
Conjugation: Cy7
Public Immunogen Range: 251-351/351
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Rabbit