Anti-Occludin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-1495R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Occludin
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Pig,Human,Mouse
- Western blot:Yes
- Epitope:431-522/522
- Form:Liquid
- Gene ID:18260
- Antigen synonyms:AI53564|Occludin|Ocl|Ocln
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:57 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from mouse Occludin
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
May play a role in the formation and regulation of the tight junction (TJ) paracellular permeability barrier.
Type: Primary
Antigen: Occludin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 431-522/522
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Pig