Anti-Nogo-A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-0134R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Nogo-A
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Epitope:301-400/1192
- Form:Liquid
- Gene ID:68585
- Antigen synonyms:Nogo-B|AA409940|NSP-CL|NgA|mKIAA0886|Kiaa0886|AA407876|AA960376|Nogo protein|mKIAA4153|Nogo-C|Neurite outgrowth inhibitor|Reticulon-4|Rtn4|NOGO|C130026I10Rik|1110020G17Rik|Nogo-A|ASY
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:131 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from mouse RTN4 protein
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Developmental neurite growth regulatory factor with a role as a negative regulator of axon-axon adhesion and growth, and as a facilitator of neurite branching. Regulates neurite fasciculation, branching and extension in the developing nervous system. Involved in down-regulation of growth, stabilisation of wiring and restriction of plasticity in the adult CNS. Regulates the radial migration of cortical neurons via an RTN4R-LINGO1 containing receptor complex. May inhibit BACE1 activity and amyloid precursor protein processing.
Type: Primary
Antigen: Nogo-A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 301-400/1192
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat