Anti-RASL10A Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-5722R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:RASL10A
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Epitope:51-150/203
- Form:Liquid
- Gene ID:10633
- Antigen synonyms:Ras like protein family member 10A|Rasl10a RAS like, family 10, member A|RSLBA_HUMAN|Ras like protein RRP22|Ras related protein on chromosome 22 homolog|RP23 64E17.6 antibody RRP22|RAS like, family 10, member A|RASL10A protein|Ras related protein on chromosome 22|RAS related on chromosome 22
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:23 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human RASL
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Ras proteins are members of a superfamily of related small GTPases. RRP22 can be posttranslationally modified by farnesyl. RRP22 inhibits cell growth and promotes caspase-independent cell death. The specific function of RASL10A is not yet known.
Type: Primary
Antigen: RASL10A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/203
Host: Rabbit
Isotype: IgG
Reactivity: Mouse