Anti-Anterior Gradient 2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-5123R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Anterior Gradient 2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:51-150/175
- Form:Liquid
- Gene ID:10551
- Antigen synonyms:AG2|HAG-2|PDIA17|AG-2|Anterior gradient protein 2 homolog|AGR2|UNQ515/PRO1030|HPC8|XAG-2|Secreted cement gland protein XAG-2 homolog|GOB-4|HEL-S-116
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:20 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Anterior Gradient 2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Required for MUC2 post-transcriptional synthesis and secretion. May play a role in the production of mucus by intestinal cells (By similarity). Proto-oncogene that may play a role in cell migration, cell differentiation and cell growth.
Type: Primary
Antigen: Anterior Gradient 2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/175
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat