Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-ADCY7 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-3924R-A680
 :  Bioss
Anti-ADCY7 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-3924R-A680
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    ADCY7
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    901-1080/1080
  • Form:
    Liquid
  • Gene ID:
    113
  • Antigen synonyms:
    Adenylyl cyclase 7|Adenylate cyclase type 7|ATP pyrophosphate-lyase 7|AC7|KIAA37|ADCY7|Adenylate cyclase type VII
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    120 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human ADCY7
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

This gene encodes a membrane-bound adenylate cyclase that catalyses the formation of cyclic AMP from ATP and is inhibitable by calcium. The product of this gene is a member of the adenylyl cyclase class-4/guanylyl cyclase enzyme family that is characterised by the presence of twelve membrane-spanning domains in its sequences.

Type: Primary
Antigen: ADCY7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 901-1080/1080
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat