Anti-Protor-1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-3866R-A680
: Bioss
- Antibody type:Primary
- Antigen name:Protor-1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:51-150/388
- Form:Liquid
- Gene ID:55615
- Antigen synonyms:AU043908|C030017C09Rik|Proline-rich protein 5|Protor-1|FLJ20185k|MGC109513|RP1-181C9.3|PTOR1|PP610|MGC32512|PRR5_HUMAN|Proline rich 5 renal|Protein observed with Rictor-1|FLJ20185|PRR5|PROTOR1|Arhgap8|C78947
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:43 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Protor-1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Protor-1 plays an important role in regulation of PDGF receptor expression and in modulation of PDGF signaling. It is thought that Protor-1 may act as a tumor suppressor in breast cancer.
Type: Primary
Antigen: Protor-1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/388
Host: Rabbit
Isotype: IgG
Reactivity: