Anti-Histone H2A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-3779R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Histone H2A
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:10-100/130
- Form:Liquid
- Gene ID:8338
- Antigen synonyms:Histone H2A/q|Histone H2A type 2-C|HIST2H2AC|H2A|H2AFQ|H2A/q|Histone H2A-GL101|H2A-GL101
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:14 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Histone H2A
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
Type: Primary
Antigen: Histone H2A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 10-100/130
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat