Anti-IKB beta Ser23 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-3191R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:IKB beta Ser23
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:4793
- Antigen synonyms:IKB beta Ser23|IkappaBbeta|IKB beta S23|Thyroid receptor-interacting protein 9|p-IKB beta Ser23|IkB-B|IKBB|IkB-beta|NFKBIBSer23|IkB-betaSer22|IKB beta|IkB beta|I-kappa-B-beta|p-IKB beta S23|NF-kappa-B inhibitor beta|TR-interacting protein 9|NFKBIBphospho S23|NFKBIB|NF-kappa-BIB|TRIP-9
- Modification:Phosphorylation (Ser23 )
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:39 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human NFKBIB around the phosphorylation site of Ser23
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
The protein encoded by this gene belongs to the NF-kappa-B inhibitor family, which inhibit NF-kappa-B by complexing with, and trapping it in the cytoplasm. Phosphorylation of serine residues on these proteins by kinases marks them for destruction via the ubiquitination pathway, thereby allowing activation of the NF-kappa-B, which translocates to the nucleus to function as a transcription factor. Alternatively spliced transcript variants have been found for this gene.
Type: Primary
Antigen: IKB beta Ser23
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human